Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

AXYTF150

Pipette filter tips

universal fit, size 150 μL, clear, pkg of 10x1000tips/cs

Synonym(s):

pipet tips

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41121601
NACRES:
NB.02
volume:
150 μL
packaging:
pkg of 10x1000tips/cs
manufacturer/tradename:
Axygen Inc TF-150
Corning
Pricing and availability is not currently available.

manufacturer/tradename

Axygen Inc TF-150
Corning

packaging

pkg of 10x1000tips/cs

size

150 μL

volume

150 μL

color

clear

Looking for similar products? Visit Product Comparison Guide

Other Notes

Browse all pipette tips

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service