AXYSPE200F9650
AxySep SPE plate
200 SPE, 96 well, fritted
Synonym(s):
Corning plate
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
41122102
material
clear plate
clear wells
sterility
non-sterile
packaging
case of 50 ea
manufacturer/tradename
Axygen Inc
Corning
technique(s)
solid phase extraction (SPE): suitable
size
96 wells
pore size
20 μm filter
Looking for similar products? Visit Product Comparison Guide
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service