Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

AXYSCOLPP

Looped screw caps

with O-rings, pink, pkg of 8x500caps/cs

Synonym(s):

Corning screw cap

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41121800
NACRES:
NB.22
Pricing and availability is not currently available.

manufacturer/tradename

Axygen Inc SCO-LP-P
Corning

packaging

pkg of 8x500caps/cs

color

pink

Looking for similar products? Visit Product Comparison Guide


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service