Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

AXYRRP200CR

Pipette filter tips, for Packard® MPII and Janus® racked

robotic, racked, capacity 200 μL, clear, pkg of 5x960tips/cs

Synonym(s):

Pipette tips for Packard® MPII and Janus®

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41121601
NACRES:
NB.02
volume:
200 μL
packaging:
pkg of 5x960tips/cs
manufacturer/tradename:
Axygen Inc RRP-200-C-R
Corning
Pricing and availability is not currently available.

manufacturer/tradename

Axygen Inc RRP-200-C-R
Corning

packaging

pkg of 5x960tips/cs

capacity

200 μL

volume

200 μL

color

clear

Looking for similar products? Visit Product Comparison Guide

General description

Product is only available in China.

Other Notes

Browse all pipette tips

Legal Information

Janus is a registered trademark of PerkinElmer Inc.
Packard is a registered trademark of PerkinElmer Inc.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service