AXYQTF300RS
Pipette filter tips for Qiagen®
capacity 250 μL, black, racked, pkg of 24x96tips/cs, sterile
Synonym(s):
pipet tips
About This Item
Corning
sterility
sterile
manufacturer/tradename
Axygen Inc QTF-300-R-S
Corning
packaging
pkg of 24x96tips/cs
capacity
250 μL
volume
250 μL
color
black
Looking for similar products? Visit Product Comparison Guide
Other Notes
Legal Information
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service