Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

AXYPKF205WBRS

Pipette filter tips for PerkinElmer®/Packard® plate track, Evolution® and Mini Track instruments, wide bore, 200 μL, clear, racked, sterile

capacity 200 μL, clear, for PerkinElmer®/Packard® plate track, wide bore, racked, pkg of 5x960tips/cs, sterile

Synonym(s):

Pipette filter tips for Packard® plate track, Evolution® and Mini Track instruments

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41121601
NACRES:
NB.02
volume:
200 μL
sterility:
sterile
packaging:
pkg of 5x960tips/cs
manufacturer/tradename:
Axygen Inc PKF-205-WB-R-S
Corning
Pricing and availability is not currently available.

sterility

sterile

manufacturer/tradename

Axygen Inc PKF-205-WB-R-S
Corning

packaging

pkg of 5x960tips/cs

capacity

200 μL

volume

200 μL

color

clear

Looking for similar products? Visit Product Comparison Guide

General description

Product is only available in China.

Other Notes

Browse all pipette tips

Legal Information

Evolution is a registered trademark of PerkinElmer Inc.
Packard is a registered trademark of PerkinElmer Inc.
PerkinElmer is a registered trademark of PerkinElmer Inc.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service