Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

AXYPK50LR

MAXYMum Recovery® pipette tips for Packard® plate track, Evolution® and Mini Track instruments

capacity 50 μL, clear, racked, pkg of 5x960tips/cs

Synonym(s):

pipet tips

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41121600
NACRES:
NB.02
volume:
50 μL
packaging:
pkg of 5x960tips/cs
manufacturer/tradename:
Axygen Inc PK-50-L-R
Corning
Pricing and availability is not currently available.

manufacturer/tradename

Axygen Inc PK-50-L-R
Corning

packaging

pkg of 5x960tips/cs

capacity

50 μL

volume

50 μL

color

clear

Looking for similar products? Visit Product Comparison Guide

Other Notes

Browse all pipette tips

Legal Information

Evolution is a registered trademark of PerkinElmer Inc.
Maxymum Recovery is a registered trademark of Corning, Inc.
Packard is a registered trademark of PerkinElmer Inc.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service