AXYPCR96SGB
PCR segmented microplate
size 96 wells, blue, pkg of 5x10plates/cs
Synonym(s):
96 multiwell plate
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
41121800
NACRES:
NB.22
material:
conical bottom wells
size:
96 wells
material
conical bottom wells
manufacturer/tradename
Axygen Inc PCR-96-SG-B
Corning
packaging
pkg of 5x10plates/cs
size
96 wells
color
blue
suitability
suitable for PCR
Looking for similar products? Visit Product Comparison Guide
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service