Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

AXYPCR96MBW

PCR microplate for The Mega Base

size 96 wells, white, pkg of 5x10plates/cs

Synonym(s):

96 multiwell plate

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41121800
NACRES:
NB.22
material:
conical bottom wells
size:
96 wells
Pricing and availability is not currently available.

material

conical bottom wells

manufacturer/tradename

Axygen Inc PCR-96-MB-W
Corning

packaging

pkg of 5x10plates/cs

size

96 wells

color

white

suitability

suitable for PCR

Looking for similar products? Visit Product Comparison Guide


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service