AXYPCR96FSG
PCR microplate, full skirt
size 96 wells, conical bottom green polypropylene wells, pkg of 5x10plates/cs
Synonym(s):
96 multiwell plate
All Photos(1)
About This Item
UNSPSC Code:
41121800
NACRES:
NB.22
material:
conical bottom green polypropylene wells
size:
96 wells
material
conical bottom green polypropylene wells
sterility
non-sterile
manufacturer/tradename
Axygen Inc PCR-96-FS-G
Corning
packaging
pkg of 5x10plates/cs
maximum volume
200 μL
size
96 wells
working volume
200 μL
suitability
suitable for PCR
binding type
non-treated surface
Choose from one of the most recent versions:
Certificates of Analysis (COA)