Key Documents
AXYPCR96BK
Corning® Axygen® PCR Microplate
size 96 wells, conical bottom black polypropylene wells, pkg of 5x10plates/cs
Synonym(s):
96 multiwell plate
About This Item
UNSPSC Code:
41121800
NACRES:
NB.22
material:
conical bottom black polypropylene wells
size:
96 wells
Pricing and availability is not currently available.
material
conical bottom black polypropylene wells
sterility
non-sterile
manufacturer/tradename
Axygen Inc PCR-96-BK
Corning
packaging
pkg of 5x10plates/cs
maximum volume
200 μL
size
96 wells
working volume
200 μL
suitability
suitable for PCR
binding type
non-treated surface
Legal Information
Axygen is a registered trademark of Corning, Inc.
Corning is a registered trademark of Corning, Inc.
Choose from one of the most recent versions:
Certificates of Analysis (COA)