Key Documents
AXYPCR384M2W
PCR microplate for ABI 3730, 3100 and MJ Thermocyclers
size 384 wells, conical bottom white polypropylene wells, pkg of 5x10plates/cs
Synonym(s):
384 multiwell plate
About This Item
UNSPSC Code:
41121800
NACRES:
NB.22
material:
conical bottom white polypropylene wells
size:
384 wells
material
conical bottom white polypropylene wells
sterility
non-sterile
manufacturer/tradename
Axygen Inc PCR-384M2-W
Corning
packaging
pkg of 5x10plates/cs
maximum volume
50 μL
size
384 wells
working volume
50 μL
suitability
suitable for PCR
binding type
non-treated surface
Choose from one of the most recent versions:
Certificates of Analysis (COA)