AXYPCR384LC480WBC
PCR microplate for Roche® 480 LightCycler®
barcoded, with film, size 384 wells, pkg of 5x10plates/cs
Synonym(s):
384 multiwell plate
About This Item
Recommended Products
material
conical bottom white polypropylene wells
sterility
non-sterile
manufacturer/tradename
Axygen Inc PCR-384-LC480-W-BC
Corning
packaging
pkg of 5x10plates/cs
maximum volume
30 μL
size
384 wells
working volume
30 μL
suitability
suitable for PCR
binding type
non-treated surface
Legal Information
Choose from one of the most recent versions:
Certificates of Analysis (COA)