Key Documents
AXYPCR38455O
Corning® Axygen® PCR Microplate
size 384 wells, conical bottom orange polypropylene wells, maximum volume 55 μL, pkg of 5x10plates/cs
Synonym(s):
384 multiwell plate
About This Item
UNSPSC Code:
41121800
NACRES:
NB.22
material:
conical bottom orange polypropylene wells
size:
384 wells
material
conical bottom orange polypropylene wells
sterility
non-sterile
manufacturer/tradename
Axygen Inc PCR-384-55-O
Corning
packaging
pkg of 5x10plates/cs
maximum volume
55 μL
size
384 wells
working volume
50 μL
suitability
suitable for (thermal cycling)
binding type
non-treated surface
Legal Information
Axygen is a registered trademark of Corning, Inc.
Corning is a registered trademark of Corning, Inc.
Choose from one of the most recent versions:
Certificates of Analysis (COA)