Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

AXYMCT175O

Microtubes

size 1.7 mL, orange, pkg of 10x500tubes/cs

Synonym(s):

centrifuge tubes

Sign Into View Organizational & Contract Pricing

Select a Size

1 PKG
$303.00

$303.00


Available to ship onMay 05, 2025Details


Notify Me

Get notified when this item is ready to ship via email.


Select a Size

Change View
1 PKG
$303.00

About This Item

UNSPSC Code:
41121702
NACRES:
NB.22

$303.00


Available to ship onMay 05, 2025Details


Notify Me

Get notified when this item is ready to ship via email.

manufacturer/tradename

Axygen Inc MCT-175-O
Corning

packaging

pkg of 10x500tubes/cs

size

1.7 mL

color

orange

Looking for similar products? Visit Product Comparison Guide


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service