Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

AXYLT384R

Pipette tips for Molecular Device Liberty

racked, capacity 30 μL, pkg of 5x3840tips/cs, clear

Synonym(s):

Pipette Tips

Sign Into View Organizational & Contract Pricing

Select a Size

1 PKG
$4,050.00

$4,050.00


Check Cart for Availability


Select a Size

Change View
1 PKG
$4,050.00

About This Item

UNSPSC Code:
41121600
NACRES:
NB.02
volume:
30 μL
packaging:
pkg of 5x3840tips/cs
manufacturer/tradename:
Axygen Inc LT-384-R
Corning

$4,050.00


Check Cart for Availability

manufacturer/tradename

Axygen Inc LT-384-R
Corning

packaging

pkg of 5x3840tips/cs

capacity

30 μL

volume

30 μL

color

clear

Looking for similar products? Visit Product Comparison Guide

Other Notes

Browse all pipette tips

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service