Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

AXYLT384BKR

Pipette tips for Molecular Device Liberty

conductive, racked, capacity 30 μL, pkg of 5x3840tips/cs, black

Synonym(s):

Pipette Tips

Sign Into View Organizational & Contract Pricing

Select a Size

1 PKG
$3,810.00

$3,810.00


Check Cart for Availability


Select a Size

Change View
1 PKG
$3,810.00

About This Item

UNSPSC Code:
41121600
NACRES:
NB.02
volume:
30 μL
sterility:
non-sterile
feature:
DNA free
RNase free
barrier: no
endotoxin free
filter: no
packaging:
pkg of 5x3840tips/cs
manufacturer/tradename:
Axygen Inc LT-384-BK-R
Corning

$3,810.00


Check Cart for Availability

material

black

sterility

non-sterile

feature

DNA free
RNase free
barrier: no
endotoxin free
filter: no

manufacturer/tradename

Axygen Inc LT-384-BK-R
Corning

packaging

pkg of 5x3840tips/cs

capacity

30 μL

volume

30 μL

color

black

Looking for similar products? Visit Product Comparison Guide

Features and Benefits

  • Black polypropylene nonpyrogenic tip
  • To ensure consistent performance and quality, tips are assembled via automation and manufactured to stringent specifications under a strict process
  • Compatible with molecular device fluorescent imaging plate reader (FLIPR) liberty 384 workstation

Other Notes

Browse all pipette tips

Legal Information

Corning is a registered trademark of Corning, Inc.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service