Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

AXYHTF1000CHTRS

Corning® Axygen® Hamilton CO-RE Style Filter Tip

tip volume 1000 μL, sterile, colorless tip, case of 2304 ea (96 Tips/Rack,24 Racks/Case)

Synonym(s):

pipet tips

Sign Into View Organizational & Contract Pricing

Select a Size

2304 EACH
$308.00

$308.00


Check Cart for Availability


Select a Size

Change View
2304 EACH
$308.00

About This Item

UNSPSC Code:
41121800

$308.00


Check Cart for Availability

sterility

sterile

packaging

case of 2304 ea (96 Tips/Rack,24 Racks/Case)

manufacturer/tradename

Corning

tip volume

1000 μL

color

colorless tip

Legal Information

Axygen is a registered trademark of Corning, Inc.
Corning is a registered trademark of Corning, Inc.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service