Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

AXYHSFUCPP

qPCR sealing film roll

W × L 78 mm × 610 m, permanent

Synonym(s):

sealing mat

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41122410
Pricing and availability is not currently available.

material

polyester

sterility

non-sterile

manufacturer/tradename

Axygen Inc
Corning

technique(s)

PCR: suitable

W × L

78 mm × 610 m

Looking for similar products? Visit Product Comparison Guide


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service