Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

AXYFS700

Freeze Seal sealing film, -80 °C to 50 °C

pkg of 10x20films/cs

Synonym(s):

sealing mat

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41122410
NACRES:
NB.22
Pricing and availability is not currently available.

form

film

manufacturer/tradename

Axygen Inc FS-700
Corning

packaging

pkg of 10x20films/cs

technique(s)

cryopreservation: suitable

General description

Product is only available in China.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service