Skip to Content
MilliporeSigma
All Photos(3)

Key Documents

AV54604

Sigma-Aldrich

Anti-FAU antibody produced in rabbit

affinity isolated antibody

Synonym(s):

Anti-FAU1, Anti-FLJ22986, Anti-Finkel-Biskis-Reilly murine sarcoma virus (FBR-MuSV) ubiquitously expressed, Anti-Fub1, Anti-Fubi, Anti-MNSFbeta, Anti-RPS30

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
$541.00

$541.00


Usually ships in 5 business days. (Orders outside of US, please allow an additional 1-2 weeks for delivery)


Select a Size

Change View
100 μL
$541.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

$541.00


Usually ships in 5 business days. (Orders outside of US, please allow an additional 1-2 weeks for delivery)

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

14 kDa

species reactivity

rat, bovine, rabbit, horse, mouse, dog, human, guinea pig

concentration

0.5 mg - 1 mg/mL

technique(s)

immunohistochemistry: suitable
western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... FAU(2197)

Immunogen

Synthetic peptide directed towards the middle region of human FAU

Application

Anti-FAU antibody produced in rabbit is suitable for western blotting at a concentration of 1μg/mL.

Biochem/physiol Actions

FAU (FBR-MuSV associated ubiquitously expressed gene) gene is the cellular counterpart of the fox sequence present in the Finkel-Biskis-Reilly murine sarcoma virus (FBR-MuSV). FAU gene is mapped on to chromosome 11 at 11q13 and is ubiquitously expressed. It encodes a fusion protein comprising ubiquitin-like protein fubi at the N terminus and ribosomal protein S30 at the C terminus. Fubi belongs to ubiquitin family whereas ribosomal protein S30 is a member of S30E family of ribosomal proteins. S30 is a component of 40S ribosomal subunit and facilitates the antimicrobial activity. FAU gene is a pro-apoptotic regulatory gene that augments basal apoptosis in human T-cell lines and 293T/17 cells.

Sequence

Synthetic peptide located within the following region: VRGQTPKVAKQEKKKKKTGRAKRRMQYNRRFVNVVPTFGKKKGPNANS

Physical form

Purified antibody supplied in 1x PBS buffer with 0.09% (w/v) sodium azide and 2% sucrose.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

10 - Combustible liquids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

K Kas et al.
Biochemical and biophysical research communications, 187(2), 927-933 (1992-09-16)
The fau gene is the cellular homolog of the fox sequence in the Finkel-Biskis-Reilly Murine Sarcoma Virus (FBR-MuSV). This virus acquired the fau sequence in its reversed transcriptional orientation. Human and mouse fau cDNA's were identified and both encode a
K Kas et al.
Genomics, 17(2), 387-392 (1993-08-01)
The FAU gene is the cellular homologue of the fox sequence in the Finkel-Biskis-Reilly murine sarcoma virus (FBR-MuSV). FAU (for FBR-MuSV associated ubiquitously expressed gene) encodes the ribosomal protein S30 fused to a ubiquitin-like protein. A cosmid clone containing the
Mark R Pickard et al.
Biochimica et biophysica acta, 1812(9), 1146-1153 (2011-05-10)
FAU, which encodes a ubiquitin-like protein (termed FUBI) with ribosomal protein S30 as a carboxy-terminal extension, has recently been identified as a pro-apoptotic regulatory gene. This activity may be mediated by Bcl-G (a pro-apoptotic member of the Bcl-2 family) which

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service