Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

AV51213

Sigma-Aldrich

Anti-UXT (AB2) antibody produced in rabbit

IgG fraction of antiserum

Synonym(s):

Anti-ART-27, Anti-Ubiquitously-expressed transcript

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
$457.00

$457.00


Usually ships in 5 business days. (Orders outside of US, please allow an additional 1-2 weeks for delivery)


Select a Size

Change View
100 μL
$457.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

$457.00


Usually ships in 5 business days. (Orders outside of US, please allow an additional 1-2 weeks for delivery)

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

IgG fraction of antiserum

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

19 kDa

species reactivity

rat, dog, rabbit, human

concentration

0.5 mg - 1 mg/mL

technique(s)

western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... UXT(8409)

Immunogen

Synthetic peptide directed towards the N terminal region of human UXT

Application

Anti-UXT (AB2) antibody produced in rabbit is suitable for western blotting at a concentration of 2.5μg/ml.

Biochem/physiol Actions

Ubiquitously-expressed, prefoldin-like chaperone (UXT; ART-27) is a cofactor that modulates the androgen receptor-dependent transcription. It reportedly has roles in prostate cancer progression and induction of apoptosis by mitochondrial aggregation or TNF-induced pathway.

Sequence

Synthetic peptide located within the following region: MATPPKRRAVEATGEKVLRYETFISDVLQRDLRKVLDHRDKVYEQLAKYL

Physical form

Purified antibody supplied in 1x PBS buffer with 0.09% (w/v) sodium azide and 2% sucrose.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

10 - Combustible liquids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Samir S Taneja et al.
The Journal of biological chemistry, 279(14), 13944-13952 (2004-01-09)
Androgen receptor trapped clone-27 (ART-27) is a newly described transcriptional coactivator that binds to the N terminus of the androgen receptor (AR). Given the vital importance of AR signaling in prostate growth and differentiation, we investigated the role of ART-27
Tijuana N Moss et al.
In vitro cellular & developmental biology. Animal, 43(3-4), 139-146 (2007-06-08)
Mitochondria are the bioenergetic and metabolic centers in eukaryotic cells and play a central role in apoptosis. Mitochondrial distribution is controlled by the microtubular cytoskeleton. The perinuclear aggregation of mitochondria is one of the characteristics associated with some types of
Yuefeng Huang et al.
Molecular biology of the cell, 22(8), 1389-1397 (2011-02-11)
Proteins that directly regulate tumor necrosis factor (TNF) signaling have critical roles in determining cell death and survival. Previously we characterized ubiquitously expressed transcript (UXT)-V2 as a novel transcriptional cofactor to regulate nuclear factor-κB in the nucleus. Here we report
Min Ji Yoon et al.
Nature communications, 12(1), 1955-1955 (2021-03-31)
p62/SQSTM1 is known to act as a key mediator in the selective autophagy of protein aggregates, or aggrephagy, by steering ubiquitinated protein aggregates towards the autophagy pathway. Here, we use a yeast two-hybrid screen to identify the prefoldin-like chaperone UXT

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service