Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

AV50235

Sigma-Aldrich

Anti-Siglec-12 (AB2) antibody produced in rabbit

affinity isolated antibody

Synonym(s):

Anti-FLJ38600, Anti-S2V, Anti-SLG, Anti-Sialic acid binding Ig-like lectin 12, Anti-Siglec-1, Anti-Siglec-12, Anti-Siglec-L1, Anti-Siglec-XII

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

63 kDa

species reactivity

human

concentration

0.5 mg - 1 mg/mL

technique(s)

western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

Immunogen

Synthetic peptide directed towards the N terminal region of human SIGLEC12

Application

Anti-Siglec-12 (AB2) antibody produced in rabbit is suitable for western blotting at a concentration of 1.0μg/ml.

Biochem/physiol Actions

Sialic acid binding Ig-like lectin 12 (Siglec-12) mediates protein-carbohydrate interactions by selectively binding to sialic acid moieties. Siglec-12 recruits SHP1 and SHP2 tyrosine phosphatases and negatively recruits macrophage signaling.

Sequence

Synthetic peptide located within the following region: LCVSVLCSFSYPQNGWTASDPVHGYWFRAGDHVSRNIPVATNNPARAVQE

Physical form

Purified antibody supplied in 1x PBS buffer with 0.09% (w/v) sodium azide and 2% sucrose.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

12 - Non Combustible Liquids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Z Yu et al.
The Journal of biological chemistry, 276(26), 23816-23824 (2001-05-01)
We describe the molecular cloning and characterization of S2V, a novel sialic acid binding immunoglobulin-like lectin. The cDNA of S2V encodes a type 1 transmembrane protein with four extracellular immunoglobulin-like (Ig-like) domains and a cytoplasmic tail bearing a typical immunoreceptor
G Foussias et al.
Biochemical and biophysical research communications, 284(4), 887-899 (2001-06-21)
The sialic acid binding immunoglobulin-like lectin (Siglec) family is a recently described member of the immunoglobulin superfamily. Within this Siglec family there exists a subgroup of molecules which bear a very high degree of homology with the molecule Siglec-3 (CD33)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service