Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

AV49445

Sigma-Aldrich

Anti-ALG1 antibody produced in rabbit

affinity isolated antibody

Synonym(s):

Anti-Asparagine-linked glycosylation 1 homolog, Anti-HMAT1, Anti-HMT-1, Anti-HMT1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

52 kDa

species reactivity

yeast, human

concentration

0.5 mg - 1 mg/mL

technique(s)

western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

Storage temp.

−20°C

Gene Information

human ... ALG1(56052)

Immunogen

Synthetic peptide directed towards the N terminal region of human ALG1

Application

Anti-DLL4 antibody produced in rabbit is suitable for western blotting at a concentration of 1.0μg/ml.

Biochem/physiol Actions

ALG1, chitobiosyldiphosphodolichol beta-mannosyltransferase, catalyzes the first mannosylation step in the biosynthesis of lipid-linked oligosaccharides. Mutations in ALG1 gene results in congenital disorder of glycosylation.

Sequence

Synthetic peptide located within the following region: VVLGDVGRSPRMQYHALSLAMHGFSVTLLGFCNSKPHDELLQNNRIQIVG

Physical form

Purified antibody supplied in 1x PBS buffer with 0.09% (w/v) sodium azide and 2% sucrose.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

12 - Non Combustible Liquids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A-K Rohlfing et al.
Gene, 534(2), 345-351 (2013-10-26)
Congenital disorders of glycosylation (CDG) are a growing group of inherited metabolic disorders where enzymatic defects in the formation or processing of glycolipids and/or glycoproteins lead to variety of different diseases. The deficiency of GDP-Man:GlcNAc2-PP-dolichol mannosyltransferase, encoded by the human
Eva Morava et al.
Pediatrics, 130(4), e1034-e1039 (2012-09-12)
Deficiency of β-1,4 mannosyltransferase (MT-1) congenital disorder of glycosylation (CDG), due to ALG1 gene mutations. Features in 9 patients reported previously consisted of prenatal growth retardation, pregnancy-induced maternal hypertension and fetal hydrops. Four patients died before 5 years of age

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service