Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

AV49035

Sigma-Aldrich

Anti-LTC4S antibody produced in rabbit

IgG fraction of antiserum

Synonym(s):

Anti-Leukotriene C4 synthase, Anti-MGC33147

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

conjugate

unconjugated

antibody form

IgG fraction of antiserum

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

16 kDa

species reactivity

dog, horse, bovine, guinea pig, human

concentration

0.5 mg - 1 mg/mL

technique(s)

western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... LTC4S(4056)

Immunogen

Synthetic peptide directed towards the N terminal region of human LTC4S

Application

Anti-LTC4S antibody produced in rabbit is suitable for western blotting at a concentration of 2.5μg/ml.

Biochem/physiol Actions

Leukotriene C4 synthase (LTC4S) is involved in the biosynthesis of cysteinyl leukotrienes that are important mediators of anaphylaxis and inflammation associated with asthma. Polymorphism in the gene encoding LTC4S is associated with aspirin-induced urticarial.

Sequence

Synthetic peptide located within the following region: MKDEVALLAAVTLLGVLLQAYFSLQVISARRAFRVSPPLTTGPPEFERVY

Physical form

Purified antibody supplied in 1x PBS buffer with 0.09% (w/v) sodium azide and 2% sucrose.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

12 - Non Combustible Liquids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Damian Niegowski et al.
PloS one, 9(5), e96763-e96763 (2014-05-09)
Leukotriene (LT) C4 synthase (LTC4S) is an integral membrane protein that catalyzes the conjugation reaction between the fatty acid LTA4 and GSH to form the pro-inflammatory LTC4, an important mediator of asthma. Mouse models of inflammatory disorders such as asthma
M Sánchez-Borges et al.
Journal of investigational allergology & clinical immunology, 19(5), 375-382 (2009-10-30)
Cysteinyl leukotriene production seems to be dysregulated in patients with hypersensitivity to aspirin and other nonsteroidal anti-inflammatory drugs (NSAIDs). However, the underlying pathogenic mechanisms of these reactions are poorly understood. Previous studies have suggested a role for the A-444C polymorphism

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service