Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

AV48911

Sigma-Aldrich

Anti-ECHDC3 antibody produced in rabbit

IgG fraction of antiserum

Synonym(s):

Anti-Enoyl coenzyme A hydratase domain containing 3, Anti-FLJ20909, Anti-RP11-401F24.3

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

conjugate

unconjugated

antibody form

IgG fraction of antiserum

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

40 kDa

species reactivity

rat, dog, mouse, bovine, human, rabbit, guinea pig, yeast

concentration

0.5 mg - 1 mg/mL

technique(s)

western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... ECHDC3(79746)

Immunogen

Synthetic peptide directed towards the N terminal region of human ECHDC3

Application

Anti-ECHDC3 antibody produced in rabbit is suitable for western blotting at a concentration of 5.0μg/ml.

Biochem/physiol Actions

Enoyl CoA hydratase domain containing 3 (ECHDC3) is a cytosolic enzyme with ethylmalonyl-CoA decarboxylase activity. The ECHDC3 gene has been observed to be differentially expressed in patients with early onset of acute coronary syndrome.

Sequence

Synthetic peptide located within the following region: SLAMLKSLQSDILHDADSNDLKVIIISAEGPVFSSGHDLKELTEEQGRDY

Physical form

Purified antibody supplied in 1x PBS buffer with 0.09% (w/v) sodium azide and 2% sucrose.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

12 - Non Combustible Liquids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Vivian N Silbiger et al.
Clinica chimica acta; international journal of clinical chemistry, 421, 184-190 (2013-03-29)
Genome-wide expression analysis using microarrays has been used as a research strategy to discovery new biomarkers and candidate genes for a number of diseases. We aim to find new biomarkers for the prediction of acute coronary syndrome (ACS) with a
Carole L Linster et al.
The Journal of biological chemistry, 286(50), 42992-43003 (2011-10-22)
A limited number of enzymes are known that play a role analogous to DNA proofreading by eliminating non-classical metabolites formed by side activities of enzymes of intermediary metabolism. Because few such "metabolite proofreading enzymes" are known, our purpose was to

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service