Skip to Content
MilliporeSigma
All Photos(3)

Key Documents

AV48548

Sigma-Aldrich

Anti-MBOAT1 antibody produced in rabbit

affinity isolated antibody

Synonym(s):

Anti-MGC44669, Anti-Membrane bound O-acyltransferase domain containing 1, Anti-OACT1, Anti-dJ434O11.1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

56 kDa

species reactivity

human, horse

concentration

0.5 mg - 1 mg/mL

technique(s)

western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

Storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... MBOAT1(154141)

General description

MBOAT1 codes for membrane bound O-acyltransferase domain containing 1 protein. It functions as an enzyme that facilitates the transfer of oleoyl-CoA to hydroxyl groups of membrane proteins. A de novo translocation in MBOAT1 has been linked to brachydactyly-syndactyly syndrome.
Rabbit Anti-MBOAT1 antibody recognizes human, mouse, and rat MBOAT1.

Immunogen

Synthetic peptide directed towards the N terminal region of human MBOAT1

Application

Rabbit Anti-MBOAT1 antibody is suitable for western blot applications at a concentration of 1μg/ml.

Biochem/physiol Actions

MBOAT1 shares structural similarity with a superfamily of membrane-bound O-acetyltransferases that transfer organic compounds, usually fatty acids (e.g., cholesterol, diacylglycerol, palmitoyl), onto hydroxyl groups of membrane-embedded targets.MBOAT1 shares structural similarity with a superfamily of membrane-bound O-acetyltransferases that transfer organic compounds, usually fatty acids (e.g., cholesterol, diacylglycerol, palmitoyl), onto hydroxyl groups of membrane-embedded targets (Dauwerse et al., 2007 [PubMed 17440500]).[supplied by OMIM]. Sequence Note: The RefSeq transcript and protein were derived from genomic sequence to make the sequence consistent with the reference genome assembly. The genomic coordinates used for the transcript record were based on alignments. PRIMARYREFSEQ_SPAN PRIMARY_IDENTIFIER PRIMARY_SPAN COMP 1-304 AL158198.14 79926-80229 c 305-450 AL158198.14 20414-20559 c 451-528 AL158198.14 18975-19052 c 529-624 AL158198.14 12010-12105 c 625-680 AL355139.10 10796-10851 c 681-735 AL355139.10 8351-8405 c 736-919 AL355139.10 6169-6352 c 920-1112 AL355139.10 4060-4252 c 1113-1216 AL008627.1 1909-2012 1217-1281 AL008627.1 5097-5161 1282-1414 AL008627.1 7441-7573 1415-1566 AL008627.1 10700-10851 1567-3275 AL008627.1 18037-19745

Sequence

Synthetic peptide located within the following region: AAEPQPSSLSYRTTGSTYLHPLSELLGIPLDQVNFVVCQLVALFAAFWFR

Physical form

Purified antibody supplied in 1x PBS buffer with 0.09% (w/v) sodium azide and 2% sucrose.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

12 - Non Combustible Liquids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Johannes G Dauwerse et al.
European journal of human genetics : EJHG, 15(7), 743-751 (2007-04-19)
Here, we report a patient with a novel brachydactyly-syndactyly syndrome and a de novo translocation 46,XY,t(4;6)(q12;p23). We mapped the breakpoint and identified genes in the breakpoint region. One of the genes on chromosome 6, the membrane-associated O-acetyl transferase gene 1

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service