AV48185
Anti-ATP5B (AB1) antibody produced in rabbit
IgG fraction of antiserum, lyophilized powder
Synonym(s):
Anti-ATP synthase, H+ transporting, mitochondrial F1 complex, β polypeptide, Anti-ATPMB, Anti-ATPSB, Anti-MGC5231
About This Item
Recommended Products
biological source
rabbit
conjugate
unconjugated
antibody form
IgG fraction of antiserum
antibody product type
primary antibodies
clone
polyclonal
form
lyophilized powder
mol wt
52 kDa
species reactivity
canine, rat, human, rabbit, horse, pig, bovine, mouse
technique(s)
immunohistochemistry: suitable
western blot: suitable
immunogen sequence
IMNVIGEPIDERGPIKTKQFAPIHAEAPEFMEMSVEQEILVTGIKVVDLL
NCBI accession no.
Storage temp.
−20°C
Gene Information
human ... ATP5B(506)
Immunogen
Physical form
Disclaimer
Not finding the right product?
Try our Product Selector Tool.
Storage Class
13 - Non Combustible Solids
wgk_germany
WGK 1
flash_point_f
Not applicable
flash_point_c
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service