Microtubule-associated protein tau (MAPT) is a neuronal protein that is involved in cellular apoptosis and microtubule organization. Alterations in MAPT have been linked to neurodegenerative disorders such as Parkinson′s and Alzheimer′s disease. Rabbit Anti-MAPT antibody recognizes human, mouse, and rat MAPT.
Immunogen
Synthetic peptide directed towards the middle region of human MAPT
Application
Rabbit Anti-MAPT antibody is suitable for western blot applications at a concentration of 0.5μg/ml.
Biochem/physiol Actions
MAPT is differentially expressed in the nervous system, depending on stage of neuronal maturation and neuron type. The mutations in the gene have been associated with several neurodegenerative disorders such as Alzheimer′s disease, Pick′s disease, frontotemporal dementia, cortico-basal degeneration and progressive supranuclear palsy.This gene encodes the microtubule-associated protein tau (MAPT) whose transcript undergoes complex, regulated alternative splicing, giving rise to several mRNA species. MAPT transcripts are differentially expressed in the nervous system, depending on stage of neuronal maturation and neuron type. MAPT gene mutations have been associated with several neurodegenerative disorders such as Alzheimer′s disease, Pick′s disease, frontotemporal dementia, cortico-basal degeneration and progressive supranuclear palsy.
Sequence
Synthetic peptide located within the following region: RGAAPPGQKGQANATRIPAKTPPAPKTPPSSGEPPKSGDRSGYSSPGSPG
Physical form
Purified antibody supplied in 1x PBS buffer with 0.09% (w/v) sodium azide and 2% sucrose.
Disclaimer
Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.
Microtubule-associated protein tau (MAPT) is a neuronal protein involved in the pathogenesis of several neurodegenerative diseases including Parkinson's Disease (PD). Glycogen synthase kinase 3 beta (GSK3B) catalyzes phosphorylation in multiple sites of tau protein. However, whether or not there is
Human molecular genetics, 23(3), 648-656 (2013-10-09)
The hallmark of Alzheimer's disease (AD) pathology is an accumulation of amyloid β (Aβ) and phosphorylated tau, which are encoded by the amyloid precursor protein (APP) and microtubule-associated protein tau (MAPT) genes, respectively. Less than 5% of all AD cases
Questions
Reviews
★★★★★ No rating value
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.