Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

AV47624

Sigma-Aldrich

Anti-TCF25 antibody produced in rabbit

IgG fraction of antiserum

Synonym(s):

Anti-FKSG26, Anti-Hulp1, Anti-KIAA1049, Anti-NULP1, Anti-PRO2620, Anti-Transcription factor 25 (basic helix-loop-helix), Anti-hKIAA1049

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
$457.00

$457.00


Usually ships in 5 business days. (Orders outside of US, please allow an additional 1-2 weeks for delivery)


Select a Size

Change View
100 μL
$457.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

$457.00


Usually ships in 5 business days. (Orders outside of US, please allow an additional 1-2 weeks for delivery)

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

IgG fraction of antiserum

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

77 kDa

species reactivity

rat, guinea pig, rabbit, horse, dog, bovine, mouse, human

concentration

0.5 mg - 1 mg/mL

technique(s)

western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... TCF25(22980)

General description

TCF25 is a transcription factor with a basic helix-loop-helix domain. It negatively regulates transcription from RNA polymerase II promoters.
Rabbit Anti-TCF25 antibody recognizes canine, human, mouse, rat, and bovine TCF25.

Immunogen

Synthetic peptide directed towards the N terminal region of human TCF25

Application

Rabbit Anti-TCF25 antibody is suitable for western blot applications at a concentration of 5μg/ml.

Biochem/physiol Actions

TCF25 acts as a transcriptional repressor. It has been shown to repress transcription of SRF in vitro and so may play a role in heart development.

Sequence

Synthetic peptide located within the following region: SGKLRKKKKKQKNKKSSTGEASENGLEDIDRILERIEDSTGLNRPGPAPL

Physical form

Purified antibody supplied in 1x PBS buffer with 0.09% (w/v) sodium azide and 2% sucrose.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

10 - Combustible liquids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service