Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

AV47434

Sigma-Aldrich

Anti-CSGALNACT1 antibody produced in rabbit

affinity isolated antibody

Synonym(s):

Anti-β4GalNAcT, Anti-FLJ11264

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

61 kDa

species reactivity

bovine, mouse, dog, rabbit, rat, guinea pig, human, horse

concentration

0.5 mg - 1 mg/mL

technique(s)

western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

glycosylation

Gene Information

General description

Chondroitin sulfate N-acetylgalactosaminyltransferase 1 (CSGALNACT1) is a Golgi membrane protein that regulates the imitation of chondroitin sulphate (CS) biosynthesis. Studies in mouse have revealed that CSGALNACT1 is responsible for the synthesis of around half of the total CS chains present in epiphyseal cartilages.
Rabbit Anti-CSGALNACT1 antibody recognizes chicken, canine, zebrafish, bovine, human, mouse, and rat CSGALNACT1.

Immunogen

Synthetic peptide directed towards the N terminal region of human CSGALNACT1

Application

Rabbit Anti-CSGALNACT1 antibody is suitable for western blot applications at a concentration of 1 μg/ml.

Biochem/physiol Actions

CSGALNACT1 is a transfers 1,4-N-acetylgalactosamine (GalNAc) from UDP-GalNAc to the non-reducing end of glucuronic acid (GlcUA). It is required for addition of the first GalNAc to the core tetrasaccharide linker and for elongation of chondroitin chains.

Sequence

Synthetic peptide located within the following region: KAEVNAGVKLATEYAAVPFDSFTLQKVYQLETGLTRHPEEKPVRKDKRDE

Physical form

Purified antibody supplied in 1x PBS buffer with 0.09% (w/v) sodium azide and 2% sucrose.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

12 - Non Combustible Liquids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Chondroitin Sulfate N-Acetylgalactosaminyltransferase 1, 2
Sato, T., et al.
Handbook of Glycosyltransferases and Related Genes?, 925-933 (2014)
Sandrine Gulberti et al.
Glycobiology, 22(4), 561-571 (2011-12-14)
Glycosaminoglycan (GAG) assembly initiates through the formation of a linkage tetrasaccharide region serving as a primer for both chondroitin sulfate (CS) and heparan sulfate (HS) chain polymerization. A possible role for sulfation of the linkage structure and of the constitutive

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service