Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

AV46845

Sigma-Aldrich

Anti-TMED3 antibody produced in rabbit

IgG fraction of antiserum

Synonym(s):

Anti-C15orf22, Anti-MGC133022, Anti-P24B, Anti-Transmembrane emp24 protein transport domain containing 3

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

conjugate

unconjugated

antibody form

IgG fraction of antiserum

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

25 kDa

species reactivity

human

concentration

0.5 mg - 1 mg/mL

technique(s)

western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... TMED3(23423)

Immunogen

Synthetic peptide directed towards the C terminal region of human TMED3

Application

Anti-TMED3 antibody produced in rabbit is suitable for western blotting at a concentration of 2.5μg/mL.

Biochem/physiol Actions

TMED3 (transmembrane emp24 protein transport domain containing 3) gene encodes for single-pass type I membrane protein that belongs to EMP24/GP25L family. It plays a crucial role in vesicular protein trafficking, mainly in the early secretory pathway as well as assists in coupled localization of TMED2 and TMED10 in the cis-Golgi network. P24 protein bind to coat proteins of COPI and COPII vesicles and may facilitate the vesicle biogenesis, cargo uptake and quality control.

Sequence

Synthetic peptide located within the following region: DRARAEDLNSRVSYWSVGETIALFVVSFSQVLLLKSFFTEKRPISRAVHS

Physical form

Purified antibody supplied in 1x PBS buffer with 0.09% (w/v) sodium azide and 2% sucrose.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

12 - Non Combustible Liquids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

G Emery et al.
Journal of cell science, 113 ( Pt 13), 2507-2516 (2000-06-15)
Recent studies show that small trans-membrane proteins of approximately 22-24 kDa (the p24 family), which are grouped into 4 sub-families by sequence homology (p23, p24, p25 and p26), are involved in the early secretory pathway. In this study, we have
Nicole Jenne et al.
The Journal of biological chemistry, 277(48), 46504-46511 (2002-09-19)
The p24 proteins belong to a highly conserved family of membrane proteins that cycle in the early secretory pathway. They bind to the coat proteins of COPI and COPII vesicles, and are proposed to be involved in vesicle biogenesis, cargo

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service