Skip to Content
MilliporeSigma
All Photos(3)

Key Documents

AV46349

Sigma-Aldrich

Anti-ANP32E antibody produced in rabbit

affinity isolated antibody

Synonym(s):

Anti-Acidic (leucine-rich) Nuclear phosphoprotein 32 family, member E, Anti-LANP-L, Anti-LANPL, Anti-MGC5350

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
$541.00

$541.00


Usually ships in 5 business days. (Orders outside of US, please allow an additional 1-2 weeks for delivery)


Select a Size

Change View
100 μL
$541.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

$541.00


Usually ships in 5 business days. (Orders outside of US, please allow an additional 1-2 weeks for delivery)

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

31 kDa

species reactivity

bovine, horse, rabbit, human, dog, rat

concentration

0.5 mg - 1 mg/mL

technique(s)

immunohistochemistry: suitable
western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... ANP32E(81611)

Immunogen

Synthetic peptide directed towards the middle region of human ANP32E

Application

Anti-ANP32E antibody produced in rabbit is suitable for western blotting at a concentration of 0.25μg/ml. It is also useful for immunohistochemistry at a concentration of 4-8μg/ml.

Biochem/physiol Actions

ANP32E [Acidic (leucine-rich) Nuclear phosphoprotein 32 family, member E] also referred to as LANP-L, LANPL or MGC5350 is a histone chaperone located on chromosome 1q22 and is primarily expressed in human peripheral blood leukocytes, colon, small intestine, prostate, thymus, spleen, skeletal muscle, liver and kidney. It facilitates the genome-wide removal of histone H2A.Z/H2AFZ from the nucleosome. It also blocks the activity of protein phosphatase 2A (PP2A) but not the protein phosphatase 1. Additionally, it may play a pivotal role in cerebellar development and synaptogenesis.

Sequence

Synthetic peptide located within the following region: EDNEAPDSEEEDDEDGDEDDEEEEENEAGPPEGYEEEEEEEEEEDEDEDE

Physical form

Purified antibody supplied in 1x PBS buffer with 0.09% (w/v) sodium azide and 2% sucrose.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

10 - Combustible liquids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

M Jiang et al.
Cytogenetic and genome research, 97(1-2), 68-71 (2002-11-20)
Leucine-rich acidic nuclear protein (LANP) is a member of the leucine-rich repeats (LRRs) superfamily. Here we report on a human homologue of LANP, encoded by the gene ANP32E (alias LANPL). The gene was cloned and identified during large-scale sequencing analysis
Arnaud Obri et al.
Nature, 505(7485), 648-653 (2014-01-28)
H2A.Z is an essential histone variant implicated in the regulation of key nuclear events. However, the metazoan chaperones responsible for H2A.Z deposition and its removal from chromatin remain unknown. Here we report the identification and characterization of the human protein

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service