Skip to Content
MilliporeSigma
All Photos(3)

Key Documents

AV46110

Sigma-Aldrich

Anti-NOLC1 antibody produced in rabbit

IgG fraction of antiserum

Synonym(s):

Anti-KIAA0035, Anti-NOPP130, Anti-NOPP140, Anti-NS5ATP13, Anti-Nucleolar and coiled-body phosphoprotein 1, Anti-P130

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
$457.00

$457.00


Usually ships in 5 business days. (Orders outside of US, please allow an additional 1-2 weeks for delivery)


Select a Size

Change View
100 μL
$457.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

$457.00


Usually ships in 5 business days. (Orders outside of US, please allow an additional 1-2 weeks for delivery)

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

IgG fraction of antiserum

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

73 kDa

species reactivity

mouse, guinea pig, horse, bovine, dog, rat, human, goat, rabbit

concentration

0.5 mg - 1 mg/mL

technique(s)

immunohistochemistry: suitable
western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... NOLC1(9221)

Immunogen

Synthetic peptide directed towards the C terminal region of human NOLC1

Application

Anti-NOLC1 antibody produced in rabbit is suitable for western blotting at a concentration of 1.25μg/ml and for immunohistochemistry at a concentration of 4-8μg/ml.

Biochem/physiol Actions

NOLC1 encodes for a phosphoprotein, nucleolar and coiled-body phosphoprotein 1 that plays a crucial role in the synthesis of rRNA and the biosynthesis of ribosomes. It also induces tumorigenesis of nasopharyngeal carcinoma (NPC) and works cooperatively with tumor protein 53 for stimulating the MDM2 promoter in NPC cells.

Sequence

Synthetic peptide located within the following region: DNSFDAKRGAAGDWGERANQVLKFTKGKSFRHEKTKKKRGSYRGGSISVQ

Physical form

Purified antibody supplied in 1x PBS buffer with 0.09% (w/v) sodium azide and 2% sucrose.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

10 - Combustible liquids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Xuefei Duan et al.
Oncology reports, 30(5), 2220-2228 (2013-08-24)
Nucleolar and coiled-body phosphoprotein 1 (NOLC1) plays an essential role in the synthesis of rRNA and the biosynthesis of ribosomes. Previous studies suggest that NOLC1 is crucial for normal cell growth, and plays a role in the regulation of tumorigenesis of
Yu-Chyi Hwang et al.
The American journal of pathology, 175(1), 342-354 (2009-06-23)
Nasopharyngeal carcinoma (NPC) is one of the most common cancers among Chinese living in South China, Singapore, and Taiwan. At present, its etiological factors are not well defined. To identify which genetic alterations might be involved in NPC pathogenesis, we

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service