Synthetic peptide directed towards the N terminal region of human LOC203547
application
Anti-LOC203547 antibody produced in rabbit is suitable for western blotting at a concentration of 5.0μg/ml and for immunohistochemistry of paraffin-embedded tissue sections at a concentration of 4-8μg/ml.
Biochem/physiol Actions
The protein encoded by LOC203547 is VMA21 vacuolar H+-ATPase homolog (VMA21), a chaperone for the assembly of lysosomal vacuolar ATPase, the major proton pump of cellular membrane in mammals. Mutations in VMA21 gene causes X-linked myopathy with excessive cell vacuolation, autophagy and tissue atrophy.
Sequence
Synthetic peptide located within the following region: MERPDKAALNALQPPEFRNESSLASTLKTLLFFTALMITVPIGLYFTTKS
Physical form
Purified antibody supplied in 1x PBS buffer with 0.09% (w/v) sodium azide and 2% sucrose.
Disclaimer
Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.
The Vma21p protein in yeast is an essential assembly chaperone for the vacuolar ATPase, the major proton pump of cellular membranes. In this issue, Ramachandran et al. (2009) report that mutations in the gene encoding the human homolog VMA21 cause
X-linked Myopathy with Excessive Autophagy (XMEA) is a childhood onset disease characterized by progressive vacuolation and atrophy of skeletal muscle. We show that XMEA is caused by hypomorphic alleles of the VMA21 gene, that VMA21 is the diverged human ortholog
Questions
Reviews
★★★★★ No rating value
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.