Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

AV43978

Sigma-Aldrich

Anti-SLC25A38 antibody produced in rabbit

affinity isolated antibody

Synonym(s):

Anti-FLJ20551, Anti-FLJ22703, Anti-Solute carrier family 25, member 38

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
$541.00

$541.00


Usually ships in 5 business days. (Orders outside of US, please allow an additional 1-2 weeks for delivery)


Select a Size

Change View
100 μL
$541.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

$541.00


Usually ships in 5 business days. (Orders outside of US, please allow an additional 1-2 weeks for delivery)

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

33 kDa

species reactivity

mouse, rabbit, sheep, pig, human, bovine, horse, dog, rat

concentration

0.5 mg - 1 mg/mL

technique(s)

immunohistochemistry: suitable
western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

Immunogen

Synthetic peptide directed towards the middle region of human SLC25A38

Application

Anti-SLC25A38 antibody produced in rabbit is suitable for western blotting at a concentration of 0.25μg/ml and for immunohistochemistry of paraffin-embedded tissue sections at a concentration of 4-8μg/ml.

Biochem/physiol Actions

SLC25A38 belongs to the mitochondrial carrier family and is essential for the biosynthesis of heme during erythropoiesis. Mutations in the gene encoding this carrier protein result in erythroblast mitochondrial iron overload, characteristic of congenital sideroblastic anemias. SLC25A38 is overexpressed in acute lymphoblastic lymphoma and may be considered as a diagnostic marker.[1]

Sequence

Synthetic peptide located within the following region: VGIYFGTLYSLKQYFLRGHPPTALESVMLGVGSRSVAGVCMSPITVIKTR

Physical form

Purified antibody supplied in 1x PBS buffer with 0.09% (w/v) sodium azide and 2% sucrose.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

10 - Combustible liquids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Huaying Chen et al.
Oncology letters, 7(5), 1422-1426 (2014-04-26)
SLC25A38 is a recently identified protein that belongs to the mitochondrial solute carrier family, SLC25. Previous studies have shown that it is a pro-apoptotic protein, which regulates intrinsic caspase-dependent apoptosis. In order to clarify the effect of SLC25A38 protein expression
Caroline Kannengiesser et al.
Haematologica, 96(6), 808-813 (2011-03-12)
Congenital sideroblastic anemias are rare disorders with several genetic causes; they are characterized by erythroblast mitochondrial iron overload, differ greatly in severity and some occur within a syndrome. The most common cause of non-syndromic, microcytic sideroblastic anemia is a defect

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service