Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

AV43367

Sigma-Aldrich

Anti-LNX1 antibody produced in rabbit

IgG fraction of antiserum

Synonym(s):

Anti-LNX, Anti-Ligand of numb-protein X 1, Anti-MPDZ, Anti-PDZRN2

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

conjugate

unconjugated

antibody form

IgG fraction of antiserum

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

70 kDa

species reactivity

dog, human, rabbit, pig, horse, bovine, rat, mouse

concentration

0.5 mg - 1 mg/mL

technique(s)

western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... LNX1(84708)

General description

Ligand of numb-protein X 1 (LNX1) is an E3 ubiquitin ligase which contains RING (Really Interesting New Gene) and PDZ (Post-synaptic density, 95 kDa, Discs large, Zona Occludens-1) domains. LNX1 may function as a signaling scaffold protein with a PDZ domain has been shown to bind KCNA4, PAK6, PLEKHG5, PKC-α1, TγK2 and PBK.

Specificity

Anti-LNX1 polyclonal antibody reacts with bovine, canine, human, mouse, and rat ligand of numb-protein X 1 proteins.

Immunogen

Synthetic peptide directed towards the C terminal region of human LNX1

Application

Anti-LNX1 polyclonal antibody is used to tag ligand of numb-protein X 1 for detection and quantitation by Western blotting and in plasma by immunohistochemical (IHC) techniques. It is used as a probe to determine the roles of ligand of numb-protein X 1 as a signaling scaffold and E3 ubiquitin ligase.

Biochem/physiol Actions

LNX1 is a membrane-bound protein that is involved in signal transduction and protein interactions. LNX1 is an E3 ubiquitin-protein ligase, which mediates ubiquitination and subsequent proteasomal degradation of proteins containing phosphotyrosine binding (PTB) domains. This protein may play an important role in tumorogenesis.This gene encodes a membrane-bound protein that is involved in signal transduction and protein interactions. The encoded product is an E3 ubiquitin-protein ligase, which mediates ubiquitination and subsequent proteasomal degradation of proteins containing phosphotyrosine binding (PTB) domains. This protein may play an important role in tumorogenesis. Alternatively spliced transcript variants encoding distinct isoforms have been described. A pseudogene, which is located on chromosome 17, has been identified for this gene.

Sequence

Synthetic peptide located within the following region: SHREWDLPIYVISVEPGGVISRDGRIKTGDILLNVDGVELTEVSRSEAVA

Physical form

Purified antibody supplied in 1x PBS buffer with 0.09% (w/v) sodium azide and 2% sucrose.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

12 - Non Combustible Liquids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service