Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

AV42218

Sigma-Aldrich

Anti-HP antibody produced in rabbit

affinity isolated antibody, lyophilized powder

Synonym(s):

Anti-Haptoglobin, Anti-Hp2-α

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

lyophilized powder

mol wt

45 kDa

species reactivity

mouse, human, pig, rabbit, canine, horse, rat

technique(s)

western blot: suitable

immunogen sequence

SALGAVIALLLWGQLFAVDSGNDVTDIADDGCPKPPEIAHGYVEHSVRYQ

NCBI accession no.

UniProt accession no.

storage temp.

−20°C

Gene Information

human ... HP(3240)

General description

Haptoglobin, which consists of two α and two β chains, is a protective plasma protein that binds hemoglobin released from erythrocytes for removal by the reticuloendothelial system.

Specificity

Anti-HP polyclonal antibody reacts with rabbit, canine, pig, human, mouse, and rat haptoglobin 2-α proteins.

Immunogen

synthetic peptide corresponding to a region of human HP with an internal ID of P24641

Application

Anti-HP polyclonal antibody is used to tag the haptoglobin 2-α for detection and quantitation by Western blotting and in plasma by immunohistochemical (IHC) techniques. It is used as a probe to determine the level of haptoglobin as an indication of erythrocyte damage and hemolytic anemia.

Physical form

Lyophilized from PBS buffer with 2% sucrose

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

13 - Non Combustible Solids

wgk_germany

WGK 2

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Arianna Mazzoli et al.
Nutrients, 13(4) (2021-05-01)
Persistence of damage induced by unhealthy diets during youth has been little addressed. Therefore, we investigated the impact of a short-term fructose-rich diet on liver metabolic activity in adolescent rats and the putative persistence of alterations after removing fructose from

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service