Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

AV41738

Sigma-Aldrich

Anti-LOR antibody produced in rabbit

IgG fraction of antiserum

Synonym(s):

Anti-Loricrin, Anti-MGC111513

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203

biological source

rabbit

conjugate

unconjugated

antibody form

IgG fraction of antiserum

antibody product type

primary antibodies

clone

polyclonal

form

lyophilized powder

mol wt

26 kDa

species reactivity

human

concentration

0.5 mg - 1 mg/mL

technique(s)

immunohistochemistry: suitable
western blot: suitable

NCBI accession no.

UniProt accession no.

Storage temp.

−20°C

Gene Information

human ... LOR(4014)

General description

Loricrin is a major component (70%) of the epidermal cornified cell envelope, a cell barrier structure, which replaces the plasma membrane in terminally differentiated epidermal (corneocytes) cells. Loricrin is a marker of epidermal cell terminal differentiation.

Specificity

Anti-LOR polyclonal antibody reacts with human, mouse, rat, and bovine loricrin proteins.

Immunogen

The immunogen for anti-LOR antibody: synthetic peptide derected towards the N terminal of human LOR

Application

Anti-LOR polyclonal antibody is used to tag loricrin for detection and quantitation by Western blotting and in plasma by immunohistochemical (IHC) techniques. It is used as a probe to identify terminally differentiated keratinocytes (corneocytes) and to study the roles of loricrin in cornification of the plasma membrane of keratinocytes.

Sequence

Synthetic peptide located within the following region: GYSGGGCGGGSSGGGGGGGIGGCGGGSGGSVKYSGGGGSSGGGSGCFSSG

Physical form

Lyophilized from PBS buffer with 2% sucrose

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

12 - Non Combustible Liquids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

F H Macdonald et al.
Oncogene, 33(32), 4132-4143 (2013-09-17)
To investigate tumour progression mechanism in transgenic mouse skin carcinogenesis, inducible PTEN ablation (Δ5PTEN(flx)) was targeted to the epidermis of mice expressing activated ras(Ha)/fos oncogenes (HK1.ras and HK1.fos). RU486-treated HK1.ras/fos-Δ5PTEN(flx) epidermis exhibited significant keratinocyte proliferation resulting in hyperplasia and proliferating
Antonio Varone et al.
Cancer research, 74(11), 3067-3075 (2014-04-02)
Alterations in the balance between different metabolic pathways used to meet cellular bioenergetic and biosynthetic demands are considered hallmarks of cancer. Optical imaging relying on endogenous fluorescence has been used as a noninvasive approach to assess tissue metabolic changes during
Kyung Ah Cheong et al.
Journal of dermatological science, 76(2), 112-119 (2014-09-23)
Hydroquinone (HQ) with or without retinoic acid (RA) is routinely used for the treatment of hyperpigmented conditions. Skin irritation is a major problem with popular depigmenting agents, resulting in postinflammatory hyperpigmentation. To examine the molecular mechanism associated with skin irritation

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service