Synthetic peptide directed towards the N terminal region of human CSDC2
Biochem/physiol Actions
CSDC2 is an RNA-binding factor which binds specifically to the very 3′UTR ends of both histone H1 and H3.3 mRNAs, encompassing the polyadenylation signal. It might play a central role in the negative regulation of histone variant synthesis in the developing brain.
Sequence
Synthetic peptide located within the following region: MTSESTSPPVVPPLHSPKSPVWPTFPFHREGSRVWERGGVPPRDLPSPLP
Physical form
Purified antibody supplied in 1x PBS buffer with 0.09% (w/v) sodium azide and 2% sucrose.
Disclaimer
Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.
Journal of cellular physiology, 234(1), 740-748 (2018-08-06)
RNA-binding proteins (RBPs) have been described for cancer cell progression and differentiation, although there is still much to learn about their mechanisms. Here, using in vivo decidualization as a model, we describe the role of RBP cold shock domain containing
Questions
Reviews
★★★★★ No rating value
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.