Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

AV40684

Sigma-Aldrich

Anti-SF3A3 antibody produced in rabbit

affinity isolated antibody

Synonym(s):

Anti-PRP9, Anti-PRPF9, Anti-SAP61, Anti-SF3a60, Anti-Splicing factor 3a, subunit 3, 60 kDa

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
$541.00

$541.00


Usually ships in 5 business days. (Orders outside of US, please allow an additional 1-2 weeks for delivery)


Select a Size

Change View
100 μL
$541.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

$541.00


Usually ships in 5 business days. (Orders outside of US, please allow an additional 1-2 weeks for delivery)

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

59 kDa

species reactivity

mouse, rabbit, bovine, human, horse, yeast, dog, rat, guinea pig

concentration

0.5 mg - 1 mg/mL

technique(s)

western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... SF3A3(10946)

General description

Splicing factor 3a, subunit 3, 60 kDa (SF3A3, PRPF9, SF3a60), a 60-kDa subunit of the splicing factor 3a complex, associates with SF3A1 and SF3A2 to form a heterotrimer which helps convert 15S U2-snRNP into the active 17S pre-mRNA splicing particle. SF3a3 has been identified as a co-repressor of constitutive androstane receptor (CAR) transcriptional activity.

Specificity

Anti-SF3A3 polyclonal antibody reacts with bovine, human, mouse, rat, chicken, and canine splicing factor 3a, subunit 3, 60 kDa proteins.

Immunogen

Synthetic peptide directed towards the middle region of human SF3A3

Application

Anti-SF3A3 polyclonal antibody is used to tag splicing factor 3a, subunit 3, 60 kDa for detection and quantitation by Western blotting and in plasma by immunohistochemical (IHC) techniques. It is used as a probe to determine the roles of splicing factor 3a, subunit 3, 60 kDa in conversion of 15S U2-snRNP into the active 17S pre-mRNA splicing particle.

Biochem/physiol Actions

SF3A3 is the subunit 3 of the splicing factor 3a protein complex. The splicing factor 3a heterotrimer includes subunits 1, 2 and 3 and is necessary for the in vitro conversion of 15S U2 snRNP into an active 17S particle that performs pre-mRNA splicing. Subunit 3 interacts with subunit 1 through its amino-terminus while the zinc finger domain of subunit 3 plays a role in its binding to the 15S U2 snRNP.This This gene encodes subunit 3 of the splicing factor 3a protein complex. The splicing factor 3a heterotrimer includes subunits 1, 2 and 3 and is necessary for the in vitro conversion of 15S U2 snRNP into an active 17S particle that performs pre-mRNA splicing. Subunit 3 interacts with subunit 1 through its amino-terminus while the zinc finger domain of subunit 3 plays a role in its binding to the 15S U2 snRNP. This gene has a pseudogene on chromosome 20. Publication Note: This RefSeq record includes a subset of the publications that are available for this gene. Please see the Entrez Gene record to access additional publications.

Sequence

Synthetic peptide located within the following region: THENVQRKQARTGEEREEEEEEQISESESEDEENEIIYNPKNLPLGWDGK

Physical form

Purified antibody supplied in 1x PBS buffer with 0.09% (w/v) sodium azide and 2% sucrose.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

10 - Combustible liquids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service