Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

AV39721

Sigma-Aldrich

Anti-MINA (AB1) antibody produced in rabbit

affinity isolated antibody

Synonym(s):

Anti-MyC induced nuclear antigen

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

lyophilized powder

mol wt

32 kDa

species reactivity

rabbit, human, horse

concentration

0.5 mg - 1 mg/mL

technique(s)

immunohistochemistry: suitable
western blot: suitable

NCBI accession no.

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... MINA(84864)

General description

MINA (MDIG) is a target of c-Myc that has been implicated in cell growth and proliferation. MINA may also be involved in lung carcinogenesis.
Rabbit Anti-MINA antibody recognizes human MINA.

Immunogen

The immunogen for anti-MINA antibody: synthetic peptide derected towards the N terminal of human MINA

Application

Rabbit Anti-MINA antibody is suitable for western blot applications at a concentration of 0.25 μg/ml and for immunohistochemistry at 4-8 μg/ml.

Sequence

Synthetic peptide located within the following region: MPKKAKPTGSGKEEGPAPCKQMKLEAAGGPSALNFDSPSSLFESLISPIK

Physical form

Lyophilized from PBS buffer with 2% sucrose

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

12 - Non Combustible Liquids

wgk_germany

WGK 2

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Makoto Tsuneoka et al.
The Journal of biological chemistry, 277(38), 35450-35459 (2002-07-02)
Myc is a ubiquitous mediator of cell proliferation and can transactivate the expression of various genes through E-box sites. Here we report a novel gene, mina53 (Myc-induced nuclear antigen with a molecular mass of 53 kDa). The mina53 gene encodes
Yadong Zhang et al.
Oncogene, 24(31), 4873-4882 (2005-05-18)
Environmental or occupational exposure to mineral dusts, mainly silica and asbestos, is associated with an increased incidence of lung inflammation, fibrosis, and/or cancer. To better understand the molecular events associated with these pulmonary diseases, we attempted to identify genes that

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service