Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

AV38732

Sigma-Aldrich

Anti-KBTBD10 (AB1) antibody produced in rabbit

IgG fraction of antiserum

Synonym(s):

Anti-Kelch repeat and BTB (POZ) domain containing 10

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
$418.00

$418.00


Usually ships in 5 business days. (Orders outside of US, please allow an additional 1-2 weeks for delivery)


Select a Size

Change View
100 μL
$418.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

$418.00


Usually ships in 5 business days. (Orders outside of US, please allow an additional 1-2 weeks for delivery)

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

IgG fraction of antiserum

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

68 kDa

species reactivity

human, rat, horse, guinea pig, mouse, rabbit, bovine

concentration

0.5 mg - 1 mg/mL

technique(s)

western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... KBTBD10(10324)

Immunogen

Synthetic peptide directed towards the N terminal region of human KBTBD10

Biochem/physiol Actions

KBTBD10 (KLHL41) belongs to the Kelch-like (KLHL) group of proteins that regulate motor function and myofibrillar function. Mutations in KLHL41 result in Nemaline Myopathy, several Mendelian diseases and cancer.

Sequence

Synthetic peptide located within the following region: MDSQRELAEELRLYQSTLLQDGLKDLLDEKKFIDCTLKAGDKSLPCHRLI

Physical form

Purified antibody supplied in 1x PBS buffer with 0.09% (w/v) sodium azide and 2% sucrose.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

10 - Combustible liquids

wgk_germany

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Caroline Jirka et al.
Human molecular genetics, 28(15), 2549-2560 (2019-04-16)
Nemaline myopathy (NM) is the most common form of congenital myopathy that results in hypotonia and muscle weakness. This disease is clinically and genetically heterogeneous, but three recently discovered genes in NM encode for members of the Kelch family of
Vandana A Gupta et al.
American journal of human genetics, 93(6), 1108-1117 (2013-11-26)
Nemaline myopathy (NM) is a rare congenital muscle disorder primarily affecting skeletal muscles that results in neonatal death in severe cases as a result of associated respiratory insufficiency. NM is thought to be a disease of sarcomeric thin filaments as
Bajinder S Dhanoa et al.
Human genomics, 7, 13-13 (2013-05-17)
The Kelch-like (KLHL) gene family encodes a group of proteins that generally possess a BTB/POZ domain, a BACK domain, and five to six Kelch motifs. BTB domains facilitate protein binding and dimerization. The BACK domain has no known function yet

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service