Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

AV38468

Sigma-Aldrich

Anti-ETV5 (AB3) antibody produced in rabbit

affinity isolated antibody, lyophilized powder

Synonym(s):

Anti-ERM, Anti-Ets variant gene 5 (Ets-related molecule)

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

lyophilized powder

mol wt

58 kDa

species reactivity

, zebrafish, bovine, chicken, human, mouse, rat, canine

technique(s)

western blot: suitable

immunogen sequence

DTDLAHDSEELFQDLSQLQEAWLAEAQVPDDEQFVPDFQSDNLVLHAPPP

NCBI accession no.

UniProt accession no.

storage temp.

−20°C

Gene Information

human ... ETV5(2119)

Immunogen

synthetic peptide corresponding to a region of human ETV5 with an internal ID of P23179

Physical form

Lyophilized from PBS buffer with 2% sucrose

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

13 - Non Combustible Solids

wgk_germany

WGK 2

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service