Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

AV38141

Sigma-Aldrich

Anti-MXD1 antibody produced in rabbit

affinity isolated antibody, lyophilized powder

Synonym(s):

Anti-MAD, Anti-MAD1, Anti-MAX dimerization protein 1, Anti-MGC104659

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

lyophilized powder

mol wt

25 kDa

species reactivity

zebrafish, mouse, rat, canine, chicken, bovine, human,

technique(s)

western blot: suitable

immunogen sequence

AAAVRMNIQMLLEAADYLERREREAEHGYASMLPYNNKDRDALKRRNKSK

NCBI accession no.

UniProt accession no.

storage temp.

−20°C

Gene Information

human ... MXD1(4084)

General description

MyC associated factor X (MAX), Myc and MAX dimerization protein 1 (Mad) are basic helix-loop-helix leucine zipper transcription factors that bind to a specific hexanucleotide element of DNA, the E-box (CACGTG). Myc and Mad form heterodimers with Max to become active regulators of transcription. Myc:Max and Mad:Max heterodimers form higher order complexes that regulate a wide range of cell processes at the level of transcription. Myc:Max and Mad:Max dimerizations are regulatory control points for many myc and mad mediated cell processes.

Specificity

Anti-MXD1 polyclonal antibody reacts with bovine, canine, zebrafish, human, mouse, rat, and chicken MAX dimerization protein 1 molecules.

Immunogen

synthetic peptide corresponding to a region of human MXD1 with an internal ID of S04807

Application

Anti-MXD1 polyclonal antibody is used to tag MAX dimerization protein 1 for detection and quantitation by Western blotting and in plasma by immunohistochemical (IHC) techniques. It is used as a probe to determine the roles of MAX dimerization protein 1 in regulation of transcription.

Physical form

Lyophilized from PBS buffer with 2% sucrose

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

13 - Non Combustible Solids

wgk_germany

WGK 2

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service