Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

AV38100

Sigma-Aldrich

Anti-GATA4 antibody produced in rabbit

affinity isolated antibody

Synonym(s):

Anti-GATA binding protein 4

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

45 kDa

species reactivity

mouse, dog, rabbit, rat, human

concentration

0.5 mg - 1 mg/mL

technique(s)

western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... GATA4(2626)

General description

The GATA zinc finger transcription factors are characterized by their ability to bind to the promoter region DNA sequence "GATA" within various genes. GATA4, prevalent in endoderm-derived organs, regulates various genes involved in embryogenesis and in myocardial differentiation and function.

The previously assigned protein identifier Q5IFM8 has been merged into P43694. Full details can be found on the UniProt database.

Specificity

Anti-GATA4 polyclonal antibody reacts with human, mouse, rat, pig, and canine GATA4 transcription factors.

Immunogen

Synthetic peptide directed towards the N terminal region of human GATA4

Application

Anti-GATA4 polyclonal antibody is used to tag GATA4 proteins for detection and quantitation by Western blotting and in cells and tissues by immunohistochemical (IHC) techniques. It is used as a probe to study the possible roles of GATA4 in the regulation of embryogenesis and differentiation primarily within endoderm-derived organs and tissues.

Biochem/physiol Actions

GATA4 is a member of the GATA family of zinc-finger transcription factors. Members of this family recognize the GATA motif which is present in the promoters of many genes. GATA4is thought to regulate genes involved in embryogenesis and in myocardial differentiation and function. Mutations in this gene have been associated with cardiac septal defects.This gene encodes a member of the GATA family of zinc-finger transcription factors. Members of this family recognize the GATA motif which is present in the promoters of many genes. This protein is thought to regulate genes involved in embryogenesis and in myocardial differentiation and function. Mutations in this gene have been associated with cardiac septal defects.

Sequence

Synthetic peptide located within the following region: MYQSLAMAANHGPPPGAYEAGGPGAFMHGAGAASSPVYVPTPRVPSSVLG

Physical form

Purified antibody supplied in 1x PBS buffer with 0.09% (w/v) sodium azide and 2% sucrose.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

12 - Non Combustible Liquids

wgk_germany

WGK 2

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service