Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

AV35765

Sigma-Aldrich

Anti-HMGB3 (AB1) antibody produced in rabbit

IgG fraction of antiserum

Synonym(s):

Anti-High-mobility group box 3

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
$457.00

$457.00


Usually ships in 5 business days. (Orders outside of US, please allow an additional 1-2 weeks for delivery)


Select a Size

Change View
100 μL
$457.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

$457.00


Usually ships in 5 business days. (Orders outside of US, please allow an additional 1-2 weeks for delivery)

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

IgG fraction of antiserum

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

23 kDa

species reactivity

horse, rabbit, human, dog, bovine

concentration

0.5 mg - 1 mg/mL

technique(s)

western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... HMGB3(3149)

Immunogen

Synthetic peptide directed towards the C terminal region of human HMGB3

Biochem/physiol Actions

HMGB3 belongs to high mobility group of proteins and contains HMG box domain that binds DNA. It plays a critical role in DNA replication, nucleosome assembly and transcription. HMGB3 reportedly regulates cell proliferation, migration and invasion of B cells and gastric cancer cells.

Sequence

Synthetic peptide located within the following region: MAKGDPKKPKGKMSAYAFFVQTCREEHKKKNPEVPVNFAEFSKKCSERWK

Physical form

Purified antibody supplied in 1x PBS buffer with 0.09% (w/v) sodium azide and 2% sucrose.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

10 - Combustible liquids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yuanyuan Xie et al.
Cancer management and research, 12, 6827-6836 (2020-08-18)
The high expression of circular RNA circEPSTI1 (hsa_circRNA_000479) has been reported to be associated with the malignant potential of ovarian cancer cells and triple-negative breast cancer cells. However, the expression profile and function of circEPSTI1 in non-small cell lung cancer
Guo Chen et al.
Journal of biochemical and molecular toxicology, 37(12), e23498-e23498 (2023-08-11)
Circular RNAs (circRNAs) have been found to be associated with the development and progression of cancers including papillary thyroid carcinoma (PTC). Circ_LDLR has been reported to be highly expressed in PTC, but its underlying mechanism of action has not been
Michael J Nemeth et al.
Blood, 102(4), 1298-1306 (2003-04-26)
Hmgb3 is a member of a family of chromatin-binding proteins that can alter DNA structure to facilitate transcription factor binding. We identified the Hmgb3 cDNA in a subtractive hybridization screen for transcripts that are preferentially expressed in hematopoietic stem cells.
Kai Yin et al.
IUBMB life, 72(11), 2508-2524 (2020-09-13)
Circular RNAs (circRNAs) are non-coding RNAs that have shown to regulate the progression of human diseases, including a variety of cancers. We aimed to investigate the function and the underlying working mechanism of circRNA matrix metallopeptidase 1 (circMMP1; hsa_circ_0024108) in
Y Gong et al.
Genetics and molecular research : GMR, 12(4), 6032-6039 (2013-12-18)
Gastric cancer is a major health problem worldwide; it is the second most common cause of cancer death in the world. Recent studies indicate that the high-mobility group (HMG) of chromosomal proteins is associated with cancer progression. However, HMGB3 has

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service