Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

AV35538

Sigma-Aldrich

Anti-NUDT9 (AB1) antibody produced in rabbit

affinity isolated antibody

Synonym(s):

Anti-Nudix (Nucleoside diphosphate linked moiety X)-type motif 9

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
$541.00

$541.00


Usually ships in 5 business days. (Orders outside of US, please allow an additional 1-2 weeks for delivery)


Select a Size

Change View
100 μL
$541.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

$541.00


Usually ships in 5 business days. (Orders outside of US, please allow an additional 1-2 weeks for delivery)

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

33 kDa

species reactivity

human, dog, mouse, pig, rabbit, rat, horse

concentration

0.5 mg - 1 mg/mL

technique(s)

immunohistochemistry: suitable
western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... NUDT9(53343)

Immunogen

Synthetic peptide directed towards the C terminal region of human NUDT9

Biochem/physiol Actions

Human ADP-ribose pyrophosphatase NUDT9 belongs to a superfamily of Nudix hydrolases that catabolize potentially toxic compounds in the cell. NUDT9 alpha protein is targeted highly specifically to mitochondria, whereas the predicted protein of the NUDT9 beta transcript, which is missing this sequence, exhibits no clear subcellular localization. Investigation of the physical and enzymatic properties of NUDT9 indicates that it is functional as a monomer, optimally active at near neutral pH, and that it requires divalent metal ions and an intact Nudix motif for enzymatic activity.

Sequence

Synthetic peptide located within the following region: LEAGDDAGKVKWVDINDKLKLYASHSQFIKLVAEKRDAHWSEDSEADCHA

Physical form

Purified antibody supplied in 1x PBS buffer with 0.09% (w/v) sodium azide and 2% sucrose.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

10 - Combustible liquids

wgk_germany

WGK 2

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service