Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

AV34839

Sigma-Aldrich

Anti-ZNF258 antibody produced in rabbit

affinity isolated antibody

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
$541.00

$541.00


Usually ships in 5 business days. (Orders outside of US, please allow an additional 1-2 weeks for delivery)


Select a Size

Change View
100 μL
$541.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

$541.00


Usually ships in 5 business days. (Orders outside of US, please allow an additional 1-2 weeks for delivery)

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

79 kDa

species reactivity

human

concentration

0.5 mg - 1 mg/mL

technique(s)

immunohistochemistry: suitable
western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... ZNF258(9204)

General description

ZNF258 (ZMYM6) codes for a zinc finger protein that contains the zinc-binding motif, MYM.
Rabbit Anti-ZNF258 antibody recognizes human and canine ZNF258.

Immunogen

Synthetic peptide directed towards the middle region of human ZNF258

Application

Rabbit Anti-ZNF258 antibody is suitable for western blot applications at a concentration of 0.5 μg/ml and IHC assays at 4-8 μg/ml.

Biochem/physiol Actions

ZNF258 is part of a novel, putative, zinc-binding motif (MYM) family which encodes proteins that maintain the repeats of the MYM motif.

Sequence

Synthetic peptide located within the following region: TPVITSVMSLAKIPATLSTGNTNSVLKGAVTKEAAKIIQDESTQEDAMKF

Physical form

Purified antibody supplied in 1x PBS buffer with 0.09% (w/v) sodium azide and 2% sucrose.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

10 - Combustible liquids

wgk_germany

WGK 2

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

D Smedley et al.
Genomics, 60(2), 244-247 (1999-09-16)
Tandem repeats of a novel, putative, zinc-binding motif (MYM) have been described within the products of two, highly homologous genes: ZNF198/RAMP/FIM and ZNF261/DXS6673E. ZNF198, mapping to 13q11-q12, was recently shown to fuse to the fibroblast growth factor receptor 1 gene

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service