Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

AV34837

Sigma-Aldrich

Anti-HSF2BP antibody produced in rabbit

IgG fraction of antiserum

Synonym(s):

Anti-Heat shock transcription factor 2 binding protein

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
$457.00

$457.00


Usually ships in 5 business days. (Orders outside of US, please allow an additional 1-2 weeks for delivery)


Select a Size

Change View
100 μL
$457.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

$457.00


Usually ships in 5 business days. (Orders outside of US, please allow an additional 1-2 weeks for delivery)

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

IgG fraction of antiserum

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

38 kDa

species reactivity

rat, dog, guinea pig, mouse, bovine, human

concentration

0.5 mg - 1 mg/mL

technique(s)

immunohistochemistry: suitable
western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... HSF2BP(11077)

General description

HSF2BP codes for a protein that binds to heat shock transcription factor 2 (HSF2) and may regulate HSF2 activity. Studies have reported that HSF2BP represses the transcriptional functions of BNC1.
Rabbit Anti-HSF2BP antibody recognizes bovine, canine, human, mouse, and rat HSF2BP.

Immunogen

Synthetic peptide directed towards the N terminal region of human HSF2BP

Application

Rabbit Anti-HSF2BP antibody is suitable for western blot (1.25 μg/ml) and IHC (4-8 μg/ml).

Biochem/physiol Actions

HSF2 binding protein (HSF2BP) associates with HSF2. The interaction occurs between the trimerization domain of HSF2 and the amino terminal hydrophilic region of HSF2BP that comprises two leucine zipper motifs. HSF2BP may therefore be involved in modulating HSF2 activation.

Sequence

Synthetic peptide located within the following region: RHMGTKEEFVKVRKKDLERLTTEVMQIRDFLPRILNGEVLESFQKLKIVE

Physical form

Purified antibody supplied in 1x PBS buffer with 0.09% (w/v) sodium azide and 2% sucrose.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

10 - Combustible liquids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yujian Wu et al.
FEBS letters, 587(14), 2099-2104 (2013-05-28)
Basonuclin (BNC1), a zinc finger transcriptional factor, is essential for mouse spermatogenesis. However, the regulatory mechanisms of BNC1 in spermatogenesis are poorly understood. In this study, we identified HSF2BP, a testis-specific binding protein of HSF2, as a binding partner of

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service